Dimethyl (methylenebis(4,1-phenylene))dicarbamate 95%

Supplier: Ambeed
Warning

A528028-100MG A528028-250MG
77116-726EA 20.16 USD
77116-726 77116-728
Dimethyl (methylenebis(4,1-phenylene))dicarbamate 95%
Dimethyl (methylenebis(4,1-phenylene))dicarbamate

Precaución: for Research Use Only

Formula: C₁₇H₁₈N₂O₄
MW: 314.33 g/mol
MDL Number: MFCD00025854
CAS Number: 7450-63-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR