Methyl-2-chloro-4-fluorobenzoate 98%

Supplier: Ambeed
Warning

Synonyms: Methyl 2-chloro-4-fluorobenzenecarboxylate, Methyl 2-chloro-4-fluorobenzoate, 2-Chloro-4-fluorobenzoic acid methyl ester

A527402-100G A527402-25G A527402-500G A527402-5G
77116-718EA 85.94 USD
77116-718 77116-720 77116-722 77116-724
Methyl-2-chloro-4-fluorobenzoate 98%
Methyl-2-chloro-4-fluorobenzoate

Precaución: for Research Use Only

Formula: C₈H₆ClFO₂
MW: 188.59 g/mol
MDL Number: MFCD00173937
CAS Number: 85953-29-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR