3-Methoxysalicyl alcohol 97%

Supplier: Ambeed
Warning

A527394-1G A527394-5G
77116-714EA 7.21 USD
77116-714 77116-716
3-Methoxysalicyl alcohol 97%
3-Methoxysalicyl alcohol

Precaución: for Research Use Only

Formula: C₈H₁₀O₃
MDL Number: MFCD00008362
CAS Number: 4383-05-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR