4-(p-Tolylthio)thieno[2,3-c]pyridine-2-carboxamide 98%

Supplier: Ambeed
Warning

Synonyms: A-205804, 4-[(4-Methylphenyl)thio]thieno[2,3-c]pyridine-2-carboxamide, A 205804

A526825-10MG A526825-1MG A526825-50MG
77116-708EA 64.72 USD
77116-708 77116-710 77116-712
4-(p-Tolylthio)thieno[2,3-c]pyridine-2-carboxamide 98%
4-(p-Tolylthio)thieno[2,3-c]pyridine-2-carboxamide

Precaución: for Research Use Only

Formula: C₁₅H₁₂N₂OS₂
MW: 300.39 g/mol
MDL Number: MFCD09038566
CAS Number: 251992-66-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR