Cyclohexanecarboxamide 97%

Supplier: Ambeed
Warning

Synonyms: Carbamoylcyclohexane, Hexahydrobenzamide

A526331-100G A526331-5G
77116-704EA 158.62 USD
77116-704 77116-706
Cyclohexanecarboxamide 97%
Cyclohexanecarboxamide

Precaución: for Research Use Only

Formula: C₇H₁₃NO
MW: 127.184 g/mol
MDL Number: MFCD00009693
CAS Number: 1122-56-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR