RG-7112 98+%

Supplier: Ambeed
Warning

Synonyms: RO5045337, RG7112, ((4S,5R)-2-(4-(tert-butyl)-2-ethoxyphenyl)-4,5-bis(4-chlorophenyl)-4,5-dimethyl-4,5-dihydro-1H-imidazol-1-yl)(4-(3-(methylsulphonyl)propyl)piperazin-1-yl)methanone

A520599-1MG A520599-5MG
77116-684EA 16.98 USD
77116-684 77116-686
RG-7112 98+%
RG-7112

Precaución: for Research Use Only

Formula: C₃₈H₄₈Cl₂N₄O₄S
MW: 727.78 g/mol
MDL Number: MFCD18206914
CAS Number: 939981-39-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR