2-Fluoro-3-methylpyridin-4-amine 97%

Supplier: Ambeed
Danger

A518405-1G A518405-250MG
77116-666EA 408.91 USD
77116-666 77116-668
2-Fluoro-3-methylpyridin-4-amine 97%
2-Fluoro-3-methylpyridin-4-amine

Precaución: for Research Use Only

Formula: C₆H₇FN₂
MW: 126.13 g/mol
MDL Number: MFCD22546940
CAS Number: 1393532-84-7
UN: 2811
ADR: 6.1 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR