Nifedipine 95%

Supplier: Ambeed
Warning

A516303-100G A516303-500G A516303-5G
77116-658EA 33.99 USD
77116-658 77116-660 77116-662 77116-664
Nifedipine 95%
Nifedipine

Precaución: for Research Use Only

Formula: C₁₇H₁₈N₂O₆
MDL Number: MFCD00057326
CAS Number: 21829-25-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR