4-Methylthiophene-2-carboxaldehyde 97%

Supplier: Ambeed
Warning

Synonyms: 2-Formyl-4-methylthiophene, 4-Methylthiophene-2-carbaldehyde

A513414-100G A513414-1G A513414-250MG A513414-25G A513414-5G
77116-638EA 467.62 USD
77116-638 77116-640 77116-642 77116-644 77116-646
4-Methylthiophene-2-carboxaldehyde 97%
4-Methylthiophene-2-carboxaldehyde

Precaución: for Research Use Only

Formula: C₆H₆OS
MW: 126.17 g/mol
MDL Number: MFCD00159561
CAS Number: 6030-36-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR