Orlistat 98%

Supplier: Ambeed
Warning

Synonyms: N-Formyl-L-leucine (1S)-1-[[(2S,3S)-3-Hexyl-4-oxo-2-oxetanyl]methyl]dodecyl Ester, (S)-(S)-1-((2S,3S)-3-Hexyl-4-oxooxetan-2-yl)tridecan-2-yl 2-formamido-4-methylpentanoate

A512085-1G A512085-5G A512085-10MG
77116-634EA 12.37 USD
77116-634 77116-636 77613-612 77686-336
Orlistat 98%
Orlistat

Precaución: for Research Use Only

Formula: C₂₉H₅₃NO₅
MW: 495.75 g/mol
MDL Number: MFCD05662360
CAS Number: 96829-58-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR