2,6-Dimethylphenylhydrazine hydrochloride 97%

Supplier: Ambeed
Warning

Synonyms: (2,6-Dimethylphenyl)hydrazine hydrochloride

A506534-100G A506534-10G A506534-25G A506534-5G
77116-596EA 122.57 USD
77116-596 77116-598 77116-600 77116-602
2,6-Dimethylphenylhydrazine hydrochloride 97%
2,6-Dimethylphenylhydrazine hydrochloride

Precaución: for Research Use Only

Formula: C₈H₁₃ClN₂
MDL Number: MFCD01861265
CAS Number: 2538-61-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR