5-(Chloromethyl)-2-((tetrahydro-2H-pyran-4-yl)oxy)pyridine 97%

Supplier: Ambeed
Danger

A506464-100MG A506464-250MG
77116-592EA 40.32 USD
77116-592 77116-594
5-(Chloromethyl)-2-((tetrahydro-2H-pyran-4-yl)oxy)pyridine 97%
5-(Chloromethyl)-2-((tetrahydro-2H-pyran-4-yl)oxy)pyridine

Precaución: for Research Use Only

Formula: C₁₁H₁₄ClNO₂
MW: 227.68 g/mol
MDL Number: MFCD09817495
CAS Number: 906352-80-5
UN: 3261
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR