3-(2-Aminoethyl)-1-boc-azetidine 95%

Supplier: Ambeed
Warning

A503471-100MG A503471-250MG A503471-1G A503471-25G
77116-588EA 41.2 USD
77116-588 77116-590 77568-976 77686-318
3-(2-Aminoethyl)-1-boc-azetidine 95%
3-(2-Aminoethyl)-1-boc-azetidine

Precaución: For research use only.

Formula: C₁₀H₂₀N₂O₂
MDL Number: MFCD09878606
CAS Number: 898271-20-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR