1-(o-Tolyl)-5-(trifluoromethyl)-1H-pyrazole-4-carboxylic acid 95%

Supplier: Ambeed
Warning

A500606-100MG A500606-1G A500606-250MG A500606-5G
77116-570EA 47.75 USD
77116-570 77116-572 77116-574 77116-576
1-(o-Tolyl)-5-(trifluoromethyl)-1H-pyrazole-4-carboxylic acid 95%
1-(o-Tolyl)-5-(trifluoromethyl)-1H-pyrazole-4-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₂H₉F₃N₂O₂
MW: 270.20 g/mol
MDL Number: MFCD08446377
CAS Number: 948293-82-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR