N-(4-Ethylbenzyl)-N-methylamine 95%

Supplier: Ambeed
Warning

Synonyms: 1-(4-Ethylphenyl)-N-methylmethanamine

A500120-1G A500120-250MG A500120-5G
77116-564EA 62.6 USD
77116-564 77116-566 77116-568
N-(4-Ethylbenzyl)-N-methylamine 95%
N-(4-Ethylbenzyl)-N-methylamine

Precaución: for Research Use Only

Formula: C₁₀H₁₅N
MW: 149.233 g/mol
MDL Number: MFCD04115409
CAS Number: 568577-84-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR