1-(Difluoromethoxy)-2-nitrobenzene 97%

Supplier: Ambeed
Warning

A499862-25G A499862-5G
77116-562EA 32.96 USD
77116-562 77686-309
1-(Difluoromethoxy)-2-nitrobenzene 97%
1-(Difluoromethoxy)-2-nitrobenzene

Precaución: for Research Use Only

Formula: C₇H₅F₂NO₃
MDL Number: MFCD00221470
CAS Number: 22225-77-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR