SC-514 98%

Supplier: Ambeed
Warning

Synonyms: 4-amino-[2,3'-bithiophene]-5-carboxamide, IKK-2 Inhibitor, SC-514

A499215-100MG A499215-10MG A499215-1MG A499215-50MG A499215-5MG
77116-542EA 93.36 USD
77116-542 77116-544 77116-546 77116-548 77116-550
SC-514 98%
SC-514

Precaución: for Research Use Only

Formula: C₉H₈N₂OS₂
MW: 224.3 g/mol
MDL Number: MFCD00206459
CAS Number: 354812-17-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR