Stearic acid 95%

Supplier: Ambeed
Warning

Synonyms: Octadecanoic acid, Octadecylic acid

A497654-1000G A497654-500G
77116-530EA 26.53 USD
77116-530 77116-532 77116-534
Stearic acid 95%
Stearic acid

Precaución: for Research Use Only

Formula: C₁₈H₃₆O₂
MW: 284.48 g/mol
MDL Number: MFCD00002752
CAS Number: 57-11-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR