[4-(Methylamino)-3-nitrophenyl]methanol 95%

Supplier: Ambeed
Warning

Synonyms: (4-(Methylamino)-3-nitrophenyl)methanol

A496287-100MG A496287-250MG
77116-522EA 15.45 USD
77116-522 77116-524
[4-(Methylamino)-3-nitrophenyl]methanol 95%
[4-(Methylamino)-3-nitrophenyl]methanol

Precaución: for Research Use Only

Formula: C₈H₁₀N₂O₃
MDL Number: MFCD16689902
CAS Number: 62347-97-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR