Tetrabromophenol blue 98%

Supplier: Ambeed
Warning

A495849-1G A495849-250MG A495849-5G
77116-516EA 30.91 USD
77116-516 77116-518 77116-520
Tetrabromophenol blue 98%
Tetrabromophenol blue

Precaución: for Research Use Only

Formula: C₁₉H₆Br₈O₅S
MDL Number: MFCD00005876
CAS Number: 4430-25-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR