4-Amino-N-methylbenzenesulfonamide 98%

Supplier: Ambeed
Warning

A491525-1G A491525-5G A491525-25G
77116-484EA 19.57 USD
77116-484 77116-486 77572-554
4-Amino-N-methylbenzenesulfonamide 98%
4-Amino-N-methylbenzenesulfonamide

Precaución: for Research Use Only

Formula: C₇H₁₀N₂O₂S
MDL Number: MFCD01683104
CAS Number: 1709-52-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR