Dichlormid 97%

Supplier: Ambeed
Warning

A489530-1000G A489530-100G A489530-25G A489530-500G A489530-5G
77116-470EA 107.16 USD
77116-470 77116-472 77116-474 77116-476 77116-478
Dichlormid 97%
Dichlormid

Precaución: for Research Use Only

Formula: C₈H₁₁Cl₂NO
MDL Number: MFCD00072464
CAS Number: 37764-25-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR