4-Chloro-2,5-difluorobenzoic acid 98%

Supplier: Ambeed
Danger

A488999-100G A488999-1G A488999-25G A488999-5G
77116-454EA 184.6 USD
77116-454 77116-456 77116-458 77116-460
4-Chloro-2,5-difluorobenzoic acid 98%
4-Chloro-2,5-difluorobenzoic acid

Precaución: for Research Use Only

Formula: C₇H₃ClF₂O₂
MW: 192.55 g/mol
MDL Number: MFCD00077480
CAS Number: 132794-07-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR