Ethyl 4-hydroxy-6-methylquinoline-3-carboxylate 98%

Supplier: Ambeed
Warning

A488524-100MG A488524-1G A488524-250MG A488524-5G
77116-446EA 20.16 USD
77116-446 77116-448 77116-450 77116-452
Ethyl 4-hydroxy-6-methylquinoline-3-carboxylate 98%
Ethyl 4-hydroxy-6-methylquinoline-3-carboxylate

Precaución: for Research Use Only

Formula: C₁₃H₁₃NO₃
MW: 231.247 g/mol
MDL Number: MFCD00087702
CAS Number: 85418-82-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR