1-Bromo-4-fluorobenzene 95%

Supplier: Ambeed
Danger

Synonyms: 4-Bromofluorobenzene

A486887-10G A486887-25G
77116-438EA 5.31 USD
77116-438 77116-440
1-Bromo-4-fluorobenzene 95%
1-Bromo-4-fluorobenzene

Precaución: for Research Use Only

Formula: C₆H₄BrF
MW: 175 g/mol
MDL Number: MFCD00000342
CAS Number: 460-00-4
UN: 1993
ADR: 3,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR