2-(4-(Allyloxy)phenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 98%

Supplier: Ambeed
Warning

A473555-1G A473555-250MG A473555-5G
77116-374EA 170.98 USD
77116-374 77116-376 77116-378
2-(4-(Allyloxy)phenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 98%
2-(4-(Allyloxy)phenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

Precaución: for Research Use Only

Formula: C₁₅H₂₁BO₃
MDL Number: MFCD20491433
CAS Number: 1415236-88-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR