DL-Cystine 98%

Supplier: Ambeed
Warning

Synonyms: (H-DL-Cys-OH)2

A471772-100G A471772-1G A471772-25G A471772-5G
77116-360EA 200.85 USD
77116-360 77116-362 77116-364 77116-366
DL-Cystine 98%
DL-Cystine

Precaución: for Research Use Only

Formula: C₆H₁₂N₂O₄S₂
MW: 240.3 g/mol
Boiling Pt: 429
Melting Pt: 227
MDL Number: MFCD00084652
CAS Number: 923-32-0
Merck Index: 13,02811

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR