3-Bromobenzenesulfonyl fluoride 98%

Supplier: Ambeed
Danger

A469251-10G A469251-250MG
77116-356EA 544.87 USD
77116-356 77116-358
3-Bromobenzenesulfonyl fluoride 98%
3-Bromobenzenesulfonyl fluoride

Precaución: for Research Use Only

Formula: C₆H₄BrFO₂S
MDL Number: MFCD22053310
CAS Number: 454-65-9
UN: 3265
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR