2,3,6-Trichloroiodobenzene 95%

Supplier: Ambeed
Warning

Synonyms: 1,2,4-trichloro-3-iodobenzene

A469225-1G A469225-250MG
77116-352EA 35.01 USD
77116-352 77116-354
2,3,6-Trichloroiodobenzene 95%
2,3,6-Trichloroiodobenzene

Precaución: for Research Use Only

Formula: C₆H₂Cl₃I
MW: 307.34 g/mol
MDL Number: MFCD13176521
CAS Number: 38411-20-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR