1-Amino-3,3-diethoxypropane 97%

Supplier: Ambeed
Danger

A468946-5G A468946-1G
77116-350EA 26.78 USD
77116-350 77686-248
1-Amino-3,3-diethoxypropane 97%
1-Amino-3,3-diethoxypropane

Precaución: for Research Use Only

Formula: C₇H₁₇NO₂
MDL Number: MFCD00142581
CAS Number: 41365-75-7
UN: 3267
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR