1-Bromo-2,4-di-tert-butylbenzene 95%

Supplier: Ambeed
Warning

A468733-100MG A468733-1G A468733-250MG A468733-5G
77116-342EA 23.34 USD
77116-342 77116-344 77116-346 77116-348
1-Bromo-2,4-di-tert-butylbenzene 95%
1-Bromo-2,4-di-tert-butylbenzene

Precaución: for Research Use Only

Formula: C₁₄H₂₁Br
MW: 269.22 g/mol
MDL Number: MFCD17012753
CAS Number: 80438-66-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR