2-Bromo-1-iodo-4-(trifluoromethoxy)benzene 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
2-Bromo-4-(trifluoromethoxy)iodobenzene
A463460-100G
A463460-1G
A463460-500G
A463460-5G
77116-310EA
114.58
USD
77116-310
77116-312
77116-314
77116-316
2-Bromo-1-iodo-4-(trifluoromethoxy)benzene 98%
2-Bromo-1-iodo-4-(trifluoromethoxy)benzene
Precaución: for Research Use Only
Formula:
C₇H₃BrF₃IO MW: 366.9018 g/mol |
MDL Number:
MFCD07369009 CAS Number: 883546-30-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...