Boc-3-(trifluoromethyl)-D-phenylalanine 97%

Supplier: Ambeed
Warning

Synonyms: Boc-D-Phe(3-CF3)-OH, Boc-D-Phe(3-CF₃)-OH

A458740-1G A458740-5G A458740-250MG
77116-282EA 314.03 USD
77116-282 77116-284 77116-286 77572-444
Boc-3-(trifluoromethyl)-D-phenylalanine 97%
Boc-3-(trifluoromethyl)-D-phenylalanine

Precaución: for Research Use Only

Formula: C₁₅H₁₈F₃NO₄
MDL Number: MFCD00672525
CAS Number: 82317-82-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR