Methyl pyruvate 98%
Supplier: Ambeed
|
Danger
|
Synonyms:
Pyruvic acid ethyl mester, 2-Ketopropionic acid methyl ester, 2-Oxopropionic acid methyl ester, Methyl 2-oxopropanoate, α-Ketopropionic acid methyl ester
A457946-25G
A457946-5G
77116-274EA
9.55
USD
77116-274
77116-276
Methyl pyruvate 98%
Methyl pyruvate
Precaución: for Research Use Only
Formula:
C₄H₆O₃ MW: 102.09 g/mol |
MDL Number:
MFCD00008754 CAS Number: 600-22-6 UN: 3272 ADR: 3,Ⅲ |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...