1-Bromo-2-chloro-4-fluoro-5-nitrobenzene 98%

Supplier: Ambeed
Warning

A457608-100MG A457608-250MG A457608-5G A457608-1G
77116-266EA 22.28 USD
77116-266 77116-268 77116-270 77686-223
1-Bromo-2-chloro-4-fluoro-5-nitrobenzene 98%
1-Bromo-2-chloro-4-fluoro-5-nitrobenzene

Precaución: for Research Use Only

Formula: C₆H₂BrClFNO₂
MW: 254.44 g/mol
MDL Number: MFCD20528320
CAS Number: 111010-08-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR