(±)-Aspartic acid 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
2-Aminobutanedioic acid, DL-Aspartic acid
A457463-1000G
A457463-100G
A457463-25G
A457463-500G
77116-258EA
31.83
USD
77116-258
77116-260
77116-262
77116-264
(±)-Aspartic acid 98%
(±)-Aspartic acid
Precaución: for Research Use Only
Formula:
C₄H₇NO₄ MW: 133.10 g/mol |
MDL Number:
MFCD00063083 CAS Number: 617-45-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...