tert-Butyl 4-methylbenzoate 98%

Supplier: Ambeed
Warning

A457420-100G A457420-1G A457420-25G A457420-5G
77116-250EA 239.77 USD
77116-250 77116-252 77116-254 77116-256
tert-Butyl 4-methylbenzoate 98%
tert-Butyl 4-methylbenzoate

Precaución: for Research Use Only

Formula: C₁₂H₁₆O₂
MW: 192.254 g/mol
MDL Number: MFCD06795961
CAS Number: 13756-42-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR