L(+)-Glutamine 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
Gln, L-Glutamine, H-Gln-OH, (S)-(+)-Glutamine, (S)-2,5-Diamino-5-oxopentanoic acid, Levoglutamide, Q
A456766-1000G
A456766-25G
A456766-500G
A456766-100G
77116-244EA
83.82
USD
77116-244
77116-246
77116-248
77594-434
L(+)-Glutamine 98%
L(+)-Glutamine
Precaución: for Research Use Only
Formula:
C₅H₁₀N₂O₃ MW: 146.15 g/mol |
MDL Number:
MFCD00008044 CAS Number: 56-85-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...