(4-(Ethylsulfinyl)phenyl)boronic acid 96%

Supplier: Ambeed
Danger

A453766-1G A453766-250MG A453766-5G
77116-226EA 150.38 USD
77116-226 77116-228 77116-230
(4-(Ethylsulfinyl)phenyl)boronic acid 96%
(4-(Ethylsulfinyl)phenyl)boronic acid

Precaución: for Research Use Only

Formula: C₈H₁₁BO₃S
MDL Number: MFCD03092934
CAS Number: 863248-21-9
UN: 1759
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR