Methyl-2-acetamidoacrylate 97%

Supplier: Ambeed
Warning

Synonyms: 2-Acetamidoacrylic Acid Methyl Ester, Methyl 2-acetamidoacrylate

A451524-100G A451524-1G A451524-25G A451524-5G
77116-202EA 555.17 USD
77116-202 77116-204 77116-206 77116-208
Methyl-2-acetamidoacrylate 97%
Methyl-2-acetamidoacrylate

Precaución: for Research Use Only

Formula: C₆H₉NO₃
MW: 143.14 g/mol
MDL Number: MFCD00013394
CAS Number: 35356-70-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR