N-Acetyl-L-tyrosine ethyl ester monohydrate 98%

Supplier: Ambeed
Warning

A448829-100G A448829-1G
77116-192EA 81.38 USD
77116-192 77686-213
N-Acetyl-L-tyrosine ethyl ester monohydrate 98%
N-Acetyl-L-tyrosine ethyl ester monohydrate

Precaución: for Research Use Only

Formula: C₁₃H₁₉NO₅
MDL Number: MFCD00149093
CAS Number: 36546-50-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR