α-Bromo-3,4-difluorotoluene 96%

Supplier: Ambeed
Danger

A448637-25G A448637-5G
77116-188EA 122.57 USD
77116-188 77116-190
α-Bromo-3,4-difluorotoluene 96%
α-Bromo-3,4-difluorotoluene

Precaución: for Research Use Only

Formula: C₇H₅BrF₂
MDL Number: MFCD00009889
CAS Number: 85118-01-0
UN: 3265
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR