2-Amino-4-methoxy-6-methylpyrimidine 98%

Supplier: Ambeed
Warning

A448461-100G A448461-25G A448461-5G
77116-182EA 107.12 USD
77116-182 77116-184 77116-186
2-Amino-4-methoxy-6-methylpyrimidine 98%
2-Amino-4-methoxy-6-methylpyrimidine

Precaución: for Research Use Only

Formula: C₆H₉N₃O
MDL Number: MFCD00059768
CAS Number: 7749-47-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR