2,3-Dichlorophenylhydrazinium chloride 98%

Supplier: Ambeed
Warning

Synonyms: 2,3-Dichlorophenylhydrazine hydrochloride, (2,3-Dichlorophenyl)hydrazine hydrochloride, 1-(2,3-Dichlorophenyl)hydrazine hydrochloride

A446796-1G A446796-5G
77116-174EA 9.55 USD
77116-174 77116-176
2,3-Dichlorophenylhydrazinium chloride 98%
2,3-Dichlorophenylhydrazinium chloride

Precaución: for Research Use Only

Formula: C₆H₇Cl₃N₂
MW: 213.49 g/mol
MDL Number: MFCD00052265
CAS Number: 21938-47-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR