3-Bromo-5-fluorobenzoyl chloride 98%

Supplier: Ambeed
Danger

A444542-1G A444542-25G A444542-5G
77116-160EA 13.8 USD
77116-160 77116-162 77116-164
3-Bromo-5-fluorobenzoyl chloride 98%
3-Bromo-5-fluorobenzoyl chloride

Precaución: for Research Use Only

Formula: C₇H₃BrClFO
MDL Number: MFCD07368699
CAS Number: 887266-90-2
UN: 3265
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR