4-Chloro-2,6-difluoropyridine 95%

Supplier: Ambeed
Danger

A444441-1G A444441-250MG
77116-156EA 686.41 USD
77116-156 77116-158
4-Chloro-2,6-difluoropyridine 95%
4-Chloro-2,6-difluoropyridine

Precaución: for Research Use Only

Formula: C₅H₂ClF₂N
MW: 149.52 g/mol
MDL Number: MFCD28657830
CAS Number: 34968-33-7
UN: 2810
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR