1-Bromo-4-(difluoromethoxy)benzene 98%

Supplier: Ambeed
Danger

Synonyms: 4-(Difluoromethoxy)bromobenzene

A442188-10G A442188-25G A442188-5G A442188-100G
77116-144EA 13.8 USD
77116-144 77116-146 77116-148 77594-380
1-Bromo-4-(difluoromethoxy)benzene 98%
1-Bromo-4-(difluoromethoxy)benzene

Precaución: for Research Use Only

Formula: C₇H₅BrF₂O
MW: 223.02 g/mol
MDL Number: MFCD00221465
CAS Number: 5905-69-1
UN: 1993
ADR: 3,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR