3-Bromo-5-(trifluoromethyl)benzenesulfonyl chloride 97%

Supplier: Ambeed
Danger

A441990-10G A441990-1G A441990-5G
77116-138EA 38.12 USD
77116-138 77116-140 77116-142
3-Bromo-5-(trifluoromethyl)benzenesulfonyl chloride 97%
3-Bromo-5-(trifluoromethyl)benzenesulphonyl chloride

Precaución: for Research Use Only

Formula: C₇H₃BrClF₃O₂S
MDL Number: MFCD03094393
CAS Number: 351003-46-8
UN: 3265
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR