3-Fluoro-o-xylene 97%

Supplier: Ambeed
Danger

A437590-1G A437590-5G
77116-128EA 7.21 USD
77116-128 77116-130
3-Fluoro-o-xylene 97%
3-Fluoro-o-xylene

Precaución: for Research Use Only

Formula: C₈H₉F
MDL Number: MFCD00000323
CAS Number: 443-82-3
UN: 1993
ADR: 3,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR