3-Quinolinol 97%

Supplier: Ambeed
Warning

A437234-100G A437234-1G A437234-250MG A437234-25G A437234-5G
77116-118EA 303.84 USD
77116-118 77116-120 77116-122 77116-124 77116-126
3-Quinolinol 97%
3-Quinolinol

Precaución: for Research Use Only

Formula: C₉H₇NO
MDL Number: MFCD00169018
CAS Number: 580-18-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR